Learn More
Medchemexpress LLC L-α-phenyl-glycine-d5 | 1246820-68-7 | 98.0% | 156.19 | C8H4D5NO2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW017406S1MG
Encompass_Preferred
L-α-Phenyl-glycine-d5 is the deuterium-labeled (d5) isotopic analogue of L-α-phenylglycine, supplied as a white to off-white solid for use as an internal standard and tracer in analytical workflows. Molecular formula C8H4D5NO2, molecular weight 156.19 g·mol-1.
  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • High purity (98.0%) for reliable analytical results.
  • Suitable as a tracer in metabolic and pharmacokinetic studies.
  • Supplied as a white to off-white solid for easy handling.
  • Available in small pack sizes appropriate for analytical use.
Catalog No. 50-003-00475