Learn More
Medchemexpress LLC L-phenylalaninamide, N-[(2R,4R,5S)-5-[[(1,1-dimethylethoxy)carbonyl]amino]-4-hydroxy-1-oxo-6-phenyl- | 292632-98-5 | 99.5% | 672.85 | 25 MG

Supplier: Medchemexpress LLC 25MGHY19369
L-685458 is a potent transition state analog (TSA) γ-secretase inhibitor (GSI) that inhibits amyloid β-protein precursor γ-secretase activity with an IC50 of 17 nM. It demonstrates high selectivity compared to other aspartyl proteases and inhibits γ-secretase-mediated cleavage of APP-C99 and Notch-100. This compound is suitable for research related to Alzheimer's disease (AD) and various cancers.
- Potent transition state analog γ-secretase inhibitor
- High selectivity against other aspartyl proteases
- Inhibits γ-secretase-mediated cleavage of APP-C99 and Notch-100
- Useful for Alzheimer's disease and cancer research
- Purity of 99.48%
- Appearance as a white to off-white solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.