Learn More
Medchemexpress LLC L-phenylalanine-13C (S)-2-amino-3-phenylpropionic acid-13C | 81201-86-7 | MFCD00084036 | 99.7% | 166.18 | C8(13)CH11NO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0215S210MG
Encompass_Preferred
L-Phenylalanine-13C is a stable-isotope labeled form of the essential amino acid L-phenylalanine supplied as a white to off-white solid for use as an internal standard, tracer, or reference material in mass spectrometry, metabolic labeling, and quantitative biochemical assays. It is highly pure and water soluble for solution preparation.
  • Stable 13C-labeled amino acid for isotopic labeling and tracing.
  • High reported purity suitable for analytical use.
  • Water solubility of 25 mg/mL (ultrasonic recommended) for easy dissolution.
  • Storage stability demonstrated for long-term frozen storage.
  • Available in small mass packages for analytical workflows.
Catalog No. 50-002-96683