Learn More
Medchemexpress LLC HY-N0215 1g Medchemexpress, L-Phenylalanine CAS:63-91-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN02151G
Encompass_Preferred
Medchemexpress, HY-N0215 1g L-Phenylalanine CAS:63-91-2 L-Phenylalanine is an amino acid, which is widely used in the production of food flavors and pharmaceuticals [1] . L-Phenylalanine binds to the α2δ subunit of voltage-dependent Ca 2+ channels , with a K i value of 980 nM [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3471