Learn More
Medchemexpress LLC L-Phenylalanine, 3-(methylsulfonyl)- | 1270093-99-6 | ≥97.0% | 243.28 | 100 MG

Supplier: Medchemexpress LLC HY23529100MG
Lifitegrast impurity 6 is an impurity of Lifitegrast. This chemical is intended for laboratory use as a drug intermediate and for the manufacture of other substances. It is provided as a white to off-white solid.
- Molecular formula: C10H13NO4S
- Molecular weight: 243.28
- Purity: ≥97.0%
- Appearance: White to off-white solid
- Target: Drug intermediate
- Identified uses: Laboratory chemicals, manufacture of substances
- Storage at room temperature: 3 years
- Storage in solvent at -80°C: 2 years
- Storage in solvent at -20°C: 1 year
- Stable under recommended storage conditions
- For research use only, not fully validated for medical applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.