Learn More
Medchemexpress LLC L-Phenylalanine, 4-fluoro- | 1132-68-9 | 183.18 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00229110G
Encompass_Preferred
p-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH). It can be used to study the regulation of this enzyme and binds to the L-leucine specific receptor of Escherichia coli.
  • Substrate for tyrosine hydroxylase (TH)
  • Used to study enzyme regulation
  • Binds to the L-leucine specific receptor of Escherichia coli
Catalog No. 50-004-48957