Learn More
Medchemexpress LLC L-phenylalanine, 4-fluoro- | 1132-68-9 | 183.18 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW002291
Encompass_Preferred
p-Fluoro-L-phenylalanine (4-Fluoro-L-phenylalanine) is a substrate for tyrosine hydroxylase (TH), used to study the enzyme's regulation. It also binds to the L-leucine specific receptor of *Escherichia coli* (KD=0.26 μM). This compound is ideal for various biochemical assays.
  • Substrate for tyrosine hydroxylase (TH)
  • Used to study enzyme regulation
  • Binds to the L-leucine specific receptor of *Escherichia coli*
  • Purity of 99.96% (HPLC)
  • Appears as a white to off-white solid
Catalog No. 50-004-49492