Learn More
Medchemexpress LLC L-phenylalanine benzyl ester hydrochloride | 2462-32-0 | 98.4% | C16H18ClNO2 | 100 G
SDP
Supplier:  Medchemexpress LLC HY79908A100G
Encompass_Preferred
L-Phenylalanine benzyl ester hydrochloride is a phenylalanine derivative recognized as a beneficial ergogenic dietary substance. It influences the secretion of anabolic hormones, supports fuel supply during exercise, enhances mental performance during stress-related tasks, and helps prevent exercise-induced muscle damage. This product is intended for research use only.
  • Phenylalanine derivative
  • Functions as an ergogenic supplement
  • Influences anabolic hormone secretion
  • Supports fuel supply during exercise
  • Enhances mental performance under stress
  • Aids in preventing exercise-induced muscle damage
  • Suitable as a reagent for peptide synthesis
  • Belongs to the amino acids and derivatives classification
Catalog No. 50-004-33293