Learn More
Medchemexpress LLC L-Phenylalanine-carboxy-13C, N-[(9H-fluoren-9-ylmethoxy)carbonyl]- | 916262-84-5 | 99.8% | C2313CH21NO4 | 5 MG

Supplier: Medchemexpress LLC HY79131S55MG
This product is L-Phenylalanine-carboxy-13C, N-[(9H-fluoren-9-ylmethoxy)carbonyl]- (Fmoc-Phe-OH-13C), a highly pure chemical compound with a purity of 99.8%. It is supplied as a white to off-white solid.
- Supplied as a white to off-white solid
- Purity of 99.8% (HPLC)
- Isotopic enrichment of 99%
- Recommended for research use only
- Stable as powder at -20°C for 3 years or 4°C for 2 years
- Stable in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.