Learn More
Medchemexpress LLC L-Phenylalanine,Indole-15N | 84344-22-9 | 205.22 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0623S101MG
Encompass_Preferred
L-Phenylalanine,Indole-15N is the 15N labeled L-Tryptophan. L-Tryptophan is an essential amino acid that is the precursor of serotonin, melatonin, and vitamin B3.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-88917