Learn More
Medchemexpress LLC L-Phenylalanine,Indole-15N | 84344-22-9 | 205.22 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0623S1050MG
Encompass_Preferred
L-Phenylalanine,Indole-15N is the 15N labeled L-Tryptophan. L-Tryptophan (Tryptophan) is an essential amino acid that is the precursor of serotonin, melatonin, and vitamin B3. This compound is for research use only. We do not sell to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process
  • Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-88633