Learn More
Medchemexpress LLC L-phenylalanine, N-[(9H-fluoren-9-ylmethoxy)carbonyl]glycyl- | 117370-45-3 | MFCD00190882 | 93.9% | 444.48 g/mol | C26H24N2O5 | 50 MG

Supplier: Medchemexpress LLC HYW29892950MG
Fmoc-Gly-Phe-OH is an Fmoc-protected dipeptide (glycine-phenylalanine) used as a building block in peptide synthesis and related biochemical research. It is supplied as a solid reagent with documented purity and storage recommendations suitable for laboratory use.
- Fmoc-protected glycine-phenylalanine dipeptide
- Useful for solid-phase and solution-phase peptide synthesis
- White to off-white solid, easy to handle
- High purity by HPLC (93.9%)
- Molecular weight 444.48 g/mol and formula C26H24N2O5
- Stable at room temperature for up to 3 years in solid form
- Storage recommendations for solutions: -80°C for long-term, -20°C for short-term
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.