Learn More
Medchemexpress LLC L-Phenylalanine, N-benzoylglycyl- | 744-59-2 | 98.1% | 326.35 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137840100MG
Encompass_Preferred
Hippuryl-L-phenylalanine serves as a substrate for carboxypeptidase, a protease enzyme linked to conditions such as obesity, epilepsy, and neurodegeneration. This compound is valuable for assessing carboxypeptidase activity.
  • Substrate of carboxypeptidase
  • Utilized for determining carboxypeptidase activity
  • For research use only
Catalog No. 50-003-49646