Learn More
Medchemexpress LLC L-phenylalanyl-L-threonine | 51352-44-4 | 95.8% | 266.29 g/mol | C13H18N2O4 | 10mg
SDP
Supplier:  Medchemexpress LLC HYW96722310MG
Encompass_Preferred
L-Phenylalanyl-L-threonine is a dipeptide containing Phenylalanine and threonine L-Phenylalanyl-L-threonine can be used to synthesize the cyclic hexapeptide Banyascyclamide A[1]
Catalog No. 50-002-68374