Learn More
Cayman Chemical L-PhenylalnIn-13C6 100mg
SDP
Supplier:  Cayman Chemical 35195100 MG
Encompass_Preferred
An internal standard for the quantification of phenylalanine by GC- or LC-MS
Catalog No. 50-291-0429