Learn More
Cayman Chemical L-PhenylephrIn 1g
SDP
Supplier:  Cayman Chemical 172051 G
Encompass_Preferred
An adrenergic α1A receptor agonist (Ki = 1.4 µM) that demonstrates selectivity against the α1B and α1C receptor subtypes (Kis = 23.9 and 47.8 µM, respectively); induces smooth muscle contractions
Catalog No. 50-296-5411