Learn More
Medchemexpress LLC L-Phenylmercapturic acid-d5 | 1331906-27-4 | 99.1% | C11H8D5NO3S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY143368S1MG
Encompass_Preferred
L-Phenylmercapturic acid-d5 is a deuterium-labeled form of L-Phenylmercapturic acid. It is commonly used as a biomarker to assess exposure to aniline compounds, such as aniline and xylene. This compound can also serve as a tracer and an internal standard for quantitative analysis using various techniques.
  • Biomarker for aniline compound exposure
  • Tracer in drug development
  • Internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuteration influences pharmacokinetic and metabolic profiles
Catalog No. 50-003-56149