Learn More
Medchemexpress LLC L-Pipecolic acid | 3105-95-1 | MFCD00064347 | 99.95% | 129.16 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01273410MM/1ML
Encompass_Preferred
L-Pipecolic acid is an oral active metabolite of Lysine that can accumulate in bodily fluids in certain inherited diseases. It promotes muscle cell health and growth by enhancing protein synthesis and plays a role in gut health. This compound shows promise for research in metabolic disorders, muscle growth disorders, and intestinal diseases.
  • Promotes muscle cell health and growth.
  • Enhances protein synthesis.
  • Plays a role in promoting gut health.
  • Increases protein synthesis in C2C12 myotubes.
  • Suppresses protein degradation and autophagy activity.
  • Improves intestinal motility.
  • Increases 5-HT levels in constipated mice models.
Catalog No. 50-004-55044