Learn More
Medchemexpress LLC L-Proline-15N | 59681-31-1 | 100.0% | 116.13 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHYY0252S2
Encompass_Preferred
L-Proline-15N is the 15N-labeled L-Proline. L-Proline is one of the twenty amino acids used in living organisms as the building blocks of proteins. It is for research use only and not sold to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-93419