Learn More
U.S. Pharmacopeia L-Proline, 147-85-3, MFCD00064318, 200mg
Supplier:  U.S. Pharmacopeia 1568506
Encompass
Molecular formula C5H9NO2, Molecular weight 115.13, Melting Point 428 - 431.6 °F (220 - 222 °C) (decomposes), Boiling Point 485.96 °F (252.2 °C) at 760 mmHg, Synonyms: Proline, 2-Pyrrolidine carboxylic acid
Catalog No. NC1583112