Learn More
Alkali Scientific L Proline, 500 G
SDP
Supplier:  Alkali Scientific LPR002CP
Encompass_Preferred
L-Proline, 500 g, is a high-quality amino acid used in protein synthesis and biochemical research. As a crucial component of collagen, proline plays a role in the stability of cellular structures and the elasticity of tissues. It is commonly used in research related to tissue engineering, protein folding, and enzyme catalysis. This product is L-form Proline, which is biologically active and used in various formulations. It is available in a 500g pack for research and industrial applications. Proline is also essential in the biosynthesis of pyrroline-5-carboxylate, an intermediate in collagen biosynthesis, making it important for the study of connective tissue.
Catalog No. 50-006-60576