Learn More
Research Products International Corp L-Proline, 500 Grams
SDP
Supplier:  Research Products International Corp P50200500.0
Encompass_Preferred
L-Proline is an amino acid that plays a crucial role in the structure and function of proteins. 
It is unique among the amino acids because its side chain forms a cyclic structure, making it an imino acid. This distinctive structure gives L-Proline special properties that contribute to its biological functions.
I-Proline is also particularly important in the synthesis of collagen, a major component of connective tissues, skin, tendons, and bones. Collagen provides structural support to various tissues and contributes to their elasticity and strength. 
Catalog No. 50-213-633