Learn More
Medchemexpress LLC L-Proline p-nitroanilide trifluoroacetate salt | 108321-19-3 | MFCD00069671 | 98.0% | 349.26 | 25 G

Supplier: Medchemexpress LLC 25GHYW011155
L-Proline p-nitroanilide TFA (P-pNA TFA) is a proline derivative that serves as a colorimetric substrate for prolyl aminopeptidase. This compound is used to measure the amidolytic activity of peptide catalysts and as a substrate for enzymes that release proline from the N-terminus of small peptides. It is intended for research use only.
- Proline derivative
- Colorimetric substrate for prolyl aminopeptidase
- White to light yellow solid
- Target: Amino acid derivatives
- Store at 4°C, sealed, away from moisture
- When in solvent, store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.