Learn More
Medchemexpress LLC L-prolyl-L-alanine | 6422-36-2 | MFCD00037875 | 98.0% | 186.21 g/mol | C8H14N2O3 | 100 MG

Supplier: Medchemexpress LLC HYW130309100MG
H-Pro-Ala-OH (L-prolyl-L-alanine) is a prolyl-alanine dipeptide supplied for laboratory research use as a biochemical reagent. It is used in assay development, analytical workflows, and peptide chemistry. The material is intended for research use only and not for human therapeutic applications.
- Intended for research use and analytical applications.
- High purity (NMR) typically 98.0% enabling reliable results.
- Molecular formula C8H14N2O3 and molecular weight 186.21 g/mol.
- Identified by CAS number 6422-36-2 for unambiguous referencing.
- Storage: protect from light; in solvent -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.