Learn More
Medchemexpress LLC L-propargylglycine | 23235-01-0 | 113.11 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0060645G
Encompass_Preferred
L-Propargylglycine, also known as (S)-2-Aminopent-4-ynoic acid, is a synthetic amino acid. It can be utilized in the synthesis of folate-conjugates and corresponding metal-chelate complexes. As a click chemistry reagent, it contains an Alkyne group and is capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Synthetic amino acid
  • Useful in synthesis of folate-conjugates
  • Enables synthesis of metal-chelate complexes
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Supports bioorthogonal chemistry
  • Offers rapid and selective reactions
  • Does not interfere with biological processes under physiological conditions
Catalog No. 50-004-46309