Learn More
Medchemexpress LLC L-Pyroglutamic acid-d5 | 1086136-22-2 | 98.6% | C5H2D5NO3 | 1 MG

Supplier: Medchemexpress LLC HY76082S1MG
L-Pyroglutamic acid-d5 is a deuterium-labeled variant of L-Pyroglutamic acid, distinguished by the incorporation of stable heavy isotopes of hydrogen (deuterium). The unlabeled counterpart is recognized as the biologically active enantiomer in humans and functions as an intermediate in glutathione metabolism. Deuteration of such molecules is a strategy employed to influence the pharmacokinetic and metabolic profiles of drugs. This compound is intended for research use only and is not sold for patient use.
- Deuterium labeled form of L-Pyroglutamic acid
- For research purposes only
- Can be used as a tracer
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
- Purity: 98.6%
- Appearance: solid, white to off-white
- Molecular weight: 134.14
- Formula: C5H2D5NO3
- CAS no.: 1086136-22-2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.