Learn More
Cayman Chemical L-PyrohomoglutamIc AcId 5g
SDP
Supplier:  Cayman Chemical 316945 G
Encompass_Preferred
An amino acid building block; has been used in the synthesis of ligands for FKBPs and HDAC inhibitors
Catalog No. 50-299-1600