Learn More
Medchemexpress LLC HY-N0642 100mg Medchemexpress, α-L-Rhamnose monohydrate CAS:6155-35-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0642100MG
Encompass_Preferred
Medchemexpress, HY-N0642 100mg α-L-Rhamnose monohydrate CAS:6155-35-7 α-L-Rhamnose monohydrate is a component of the plant cell wall pectic polysaccharides rhamnogalacturonan I and rhamnogalacturonan II. α-L-Rhamnose monohydrate is also a component of bacterial polysaccharides where it plays an important role in pathogenicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3653