Learn More
Medchemexpress LLC Α-L-Rhamnose monohydrate | 6155-35-7 | 182.17 | 50 G
SDP
Supplier:  Medchemexpress LLC HYN064250G
Encompass_Preferred
α-L-Rhamnose monohydrate is a fundamental component found in the pectic polysaccharides of plant cell walls, specifically rhamnogalacturonan I and rhamnogalacturonan II. It also plays a significant role in the structure of bacterial polysaccharides, contributing to their pathogenicity.
  • Component of plant cell wall pectic polysaccharides
  • Plays an important role in bacterial pathogenicity
Catalog No. 50-003-89071