Learn More
Medchemexpress LLC L-selectin/CD62L protein, mouse (HEK293, His) | ≥98.0% | 10 UG
SDP
Supplier:  Medchemexpress LLC HYP7476610UG
Encompass_Preferred
Recombinant mouse L-selectin (CD62L) extracellular domain produced in HEK293 cells with a C-terminal His tag. L-selectin is a calcium-dependent lectin that mediates lymphocyte adhesion to endothelial cells; this purified recombinant protein is provided for immunology, cell-adhesion, and receptor-ligand interaction studies. Certificate of Analysis reports high purity.
Catalog No. 50-002-94521