Learn More
Medchemexpress LLC L-Serine-13C3,15N | 202407-34-9 | 109.06 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0650S5
Encompass_Preferred
L-Serine-13C3,15N is the 13C- and 15N-labeled L-Serine. L-Serine ((-)-Serine; (S)-Serine), one of the so-called non-essential amino acids, plays a central role in cellular proliferation. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89446