Learn More
Medchemexpress LLC L-Serine | 56-45-1 | 105.09 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN065010MM/1ML
Encompass_Preferred
L-Serine, also known as (-)-Serine or (S)-Serine, is a non-essential amino acid that plays a central role in cellular proliferation. It is intended for use as a laboratory chemical and in the manufacture of substances, strictly for research purposes and handled by qualified personnel in appropriately equipped facilities. The product has not been fully validated for medical applications.
  • Physical state: Solid
  • Stable under recommended storage conditions
  • Store at room temperature for 3 years
  • When in solvent, store at -80°C for 2 years or -20°C for 1 year
  • Predominant source of one-carbon groups for the de novo synthesis of purine nucleotides and deoxythymidine monophosphate
  • Conditionally essential amino acid in cell cultures, necessary for specific functions in the central nervous system
Catalog No. 50-003-88815