Learn More
Medchemexpress LLC L-Serine | 56-45-1 | MFCD00064224 | 105.09 | 50 MG

Supplier: Medchemexpress LLC HYN0650R50MG
L-Serine is an analytical standard and a non-essential amino acid with a central role in cellular proliferation. It is intended for research and analytical applications.
- Intended for research and analytical applications.
- Used as a reference standard.
- Commonly used in qualitative, quantitative, and methodological research experiments.
- Can be used in HPLC, GC, and MS.
- Assay of 98.37%.
- Solid appearance.
- White to off-white color.
- Molecular weight of 105.09.
- Formula C3H7NO3.
- CAS No. 56-45-1.
- Structure classification as amino acid.
- Initial source as endogenous metabolite.
- Associated with disease markers for endocrine diseases, nervous system disorders, and cardiovascular system disorders.
- Also sourced from microorganisms.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.