Learn More
Medchemexpress LLC L-Serine-d7 | 935275-35-7 | 112.14 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0650S11100MG
Encompass_Preferred
L-Serine-d7 is the deuterium-labeled form of L-Serine. L-Serine, a non-essential amino acid, plays a central role in cellular proliferation.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of elements like hydrogen and carbon.
  • Deuterium substitution has potential to affect pharmacokinetic and metabolic profiles of drugs.
  • High purity of 99.97%.
Catalog No. 50-003-88680