Learn More
Medchemexpress LLC L-serine, N-[(1,1-dimethylethoxy)carbonyl]-N-methyl- | 101772-29-6 | MFCD00037249 | 98.0% | 219.24 g/mol | C9H17NO5 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0140481G
Encompass_Preferred
Boc-N-Me-Ser-OH is the tert-butoxycarbonyl (Boc) protected N-methyl derivative of L-serine supplied as a research-grade amino acid derivative. It serves as a protected building block for introducing N-methyl serine residues in peptide synthesis, medicinal chemistry, and organic synthesis workflows.
  • Boc-protected amine for temporary protection during synthesis.
  • N-methylated serine residue useful for peptide modification.
  • High reported purity: 98.0% (NMR).
  • Molecular weight 219.24 g/mol and formula C9H17NO5.
  • Suitable for solid-phase and solution-phase peptide synthesis.
  • Supplied in small research pack sizes for laboratory use.
Catalog No. 50-003-02087