Learn More
Medchemexpress LLC L-serine, N-[(9H-fluoren-9-ylmethoxy)carbonyl]-O-(α-D-GalNAc(Ac)3)-OH | 120173-57-1 | 99.0% | 656.63 g/mol | C32H36N2O13 | 1 ML

Supplier: Medchemexpress LLC HY10400410MM 1ML
Fmoc-Ser(O-α-D-GalNAc(OAc)3)-OH is an Fmoc-protected O-glycosylated serine derivative used as a building block for synthesizing glycopeptides and glyco-conjugates. It is supplied in high-purity solid forms and as a ready-to-use 10 mM solution in DMSO, suitable for peptide synthesis, vaccine antigen assembly, and structure-activity studies.
- Purity 99.0%.
- Molecular weight 656.63 g/mol.
- Chemical formula C32H36N2O13.
- Available as solids (5-100 mg) and 10 mM solution in DMSO.
- Soluble in DMSO ≥100 mg/mL.
- Suitable for glycopeptide synthesis and vaccine antigen assembly.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.