Learn More
Medchemexpress LLC L-serine, O-acetyl-, hydrochloride | 66638-22-0 | MFCD00060169 | >97.0% | 183.59 g/mol | C5H10ClNO4 | 1 G
SDP
Supplier:  Medchemexpress LLC HY101409A1G
Encompass_Preferred
O-Acetyl-L-serine hydrochloride is the hydrochloride salt of O-acetyl-L-serine, an endogenous metabolite used as a biochemical substrate in laboratory research. It is commonly used in enzymatic and metabolic studies, including as a substrate for cysteine synthase. The compound is supplied as a solid with a defined purity suitable for analytical and preparative applications.
  • Endogenous metabolite used in biochemical research.
  • Substrate for cysteine synthase and related enzymatic assays.
  • Supplied as a solid suitable for storage and handling in laboratory settings.
  • Typical purity ≥97.0% (HPLC), supporting analytical applications.
  • Molecular weight 183.59 g/mol; molecular formula C5H10ClNO4.
Catalog No. 50-002-49128