Learn More
Medchemexpress LLC L-Serine1-13C,15N | 2483830-04-0 | 107.08 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0650S65MG
Encompass_Preferred
L-Serine1-13C,15N is the 13C- and 15N-labeled L-Serine. L-Serine ((-)-Serine; (S)-Serine), one of the so-called non-essential amino acids, plays a central role in cellular proliferation.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-89003