Learn More
Alkali Scientific L-Shape Spreader, 38 x 156mm, Individually Wrapped, 500/cs
SDP
Supplier:  Alkali Scientific TS5420
Encompass_Preferred
These L-shape spreaders are ideal for spreading bacteriological cultures evenly. The shape allows the user to rotate and apply easily throughout an entire round plate or dish. They are made of ABS and have a slight upward curve at the edge to prevent dragging or scraping.
Catalog No. 50-233-5976