Learn More
Grainger L-Shaped Workbench Laminate 72 W 30 D
Supplier:  Grainger 16Y934
Encompass_Preferred
The Pro-Line L-Shaped Workbench from Grainger is a laminate workbench with an L-shaped design. It measures 72" wide by 30" deep and features a 1-1/4" thick work surface with maple laminate finish. The workbench has steel legs with leveling feet, providing a 350 lb weight capacity. Other details include T-mold edging, scratch resistant surface, and rear access port for wire management. This commercial-grade workbench offers a spacious and durable workspace.
Catalog No. 50-276-7443