Learn More
Selleck Chemical LLC L-Sulforaphane E4007-25MG
SDP
Supplier:  Selleck Chemical LLC E400725MG
Encompass_Preferred
L-Sulforaphane ((R)-Sulforaphane) is a highly potent inducer of the Keap1/Nrf2/ARE pathway It is a more potent inducer of the carcinogen-detoxifying enzyme systems in rat liver and lung than the S-isomer
Catalog No. 50-000-42278