Learn More
Selleck Chemical LLC L-Thyroxine sodium salt pentahydrate S5193-25mg
SDP
Supplier:  Selleck Chemical LLC S519325MG
Encompass_Preferred
L-Thyroxine (Levothyroxine L-T4) sodium salt pentahydrate is a synthetic T4 hormone used to counterbalance hypothyroidism L-Thyroxine sodium salt pentahydrate can be used to induce animal models of Hyperthyroidism
Catalog No. 50-311-2257