Learn More
Cayman Chemical L-TetrahydropalmatIn 50mg
SDP
Supplier:  Cayman Chemical 2053550 MG
Encompass_Preferred
An alkaloid that antagonizes the supraspinal dopamine 2 receptor and reduces stress-induced anxiety in rats
Catalog No. 50-298-1715