Learn More
Medchemexpress LLC L-Theanine-d5 | 1217451-85-8 | MFCD09842666 | C7H9D5N2O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY15121S5MG
Encompass_Preferred
L-Theanine-d5 is a deuterium-labeled form of L-Theanine, a non-protein amino acid found in green tea leaves. It functions by blocking the binding of L-glutamic acid to glutamate receptors in the brain, demonstrating neuroprotective and anti-oxidative activities. Oral intake of L-Theanine has been observed to produce anti-stress effects through the inhibition of cortical neuron excitation. Deuterium substitution in drug molecules allows for its use as a tracer for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
  • Exhibits neuroprotective and anti-oxidative activities
  • Inhibits cortical neuron excitation for anti-stress effects
Catalog No. 50-003-75808