Learn More
Medchemexpress LLC L-Theanine-d5 (L-Glutamic Acid γ-ethyl amide-d5) | 1217451-85-8 | C7H9D5N2O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY15121S1MG
Encompass_Preferred
L-Theanine-d5 is a deuterium-labeled version of L-Theanine, a non-protein amino acid found in green tea leaves. It operates by blocking the binding of L-glutamic acid to glutamate receptors in the brain, exhibiting neuroprotective and anti-oxidative activities. Oral intake can induce anti-stress effects by inhibiting cortical neuron excitation.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Suitable for techniques such as NMR, GC-MS, or LC-MS
  • Deuterium incorporation assists in quantitation during drug development
Catalog No. 50-003-74798