Learn More
Alkali Scientific L Threonine, 100 G
SDP
Supplier:  Alkali Scientific LTH001CP
Encompass_Preferred
L-Threonine, 100 g, is an essential amino acid that plays a vital role in protein synthesis and metabolism. It is used in the formation of proteins, particularly in muscle growth and immune system support. Threonine is also involved in the production of glycine and serine, important for the synthesis of various proteins in the body. This 100g pack of L-Threonine is of high purity, making it suitable for biochemical and pharmaceutical research. It is commonly used in animal feed, pharmaceutical formulations, and as a supplement for improving protein balance in both human and animal health studies.
Catalog No. 50-006-61103