Learn More
Medchemexpress LLC L-Threonine-13C4,15N | 202468-39-1 | 124.08 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0658S35MG
Encompass_Preferred
L-Threonine-13C4,15N is the 13C- and 15N-labeled L-Threonine. L-Threonine is a natural amino acid that can be produced by microbial fermentation and is utilized in food, medicine, or feed. It is intended for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-88667