Learn More
Medchemexpress LLC L-threonine-13C4 | 55443-53-3 | MFCD00144681 | 98.0% | 123.09 g/mol | 13C4H9NO3 | 10 MG

Supplier: Medchemexpress LLC HYN0658S610MG
L-Threonine-13C4 is the 13C-labeled form of the amino acid L-threonine, supplied as a white to off-white solid for research use as a tracer or internal standard in NMR, GC-MS, and LC-MS applications.
- Purity: 98.0%.
- Isotopic labeling: 13C4.
- Molecular formula: 13C4H9NO3.
- Molecular weight: 123.09 g/mol.
- CAS number: 55443-53-3.
- Supplied quantity: 10 mg, solid.
- Appearance: white to off-white solid.
- Recommended storage: 4°C sealed; in solvent store at -80°C for up to 6 months or -20°C for 1 month.
- Typical applications: tracer or internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.