Learn More
Medchemexpress LLC L-Threonine-15N | 80681-09-0 | 120.11 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0658S5MG
Encompass_Preferred
L-Threonine-15N is the 15N-labeled L-Threonine, a natural amino acid that can be produced by microbial fermentation. It is used in food, medicine, or feed. This compound is for research use only and not sold to patients. It can be utilized as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-88747