Learn More
Medchemexpress LLC L-Threonine (Standard) | 72-19-5 | 119.12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0658R50MG
Encompass_Preferred
L-Threonine (Standard) is the analytical standard of L-Threonine. This product is intended for research and analytical applications. L-Threonine is a natural amino acid, which can be produced by microbial fermentation and is used in food, medicine, or feed. The compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard for L-Threonine
  • Intended for research and analytical applications
  • Natural amino acid
  • Can be produced by microbial fermentation
  • Used in food, medicine, or feed
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-003-88391